Anti-H3K9me1 Polyclonal Antibody
ABNOPAB14049
: Abnova
- Antibody type:Primary
- Antigen symbol:H3K9me1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Format:Liquid
- Form:Serum
- Storage buffer:0,05% sodium azide
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to region of histone H3 containing the monomethylated lysine 9 (H3K9me1).
- Tested applications:Dot Blot
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of H3K9me1.
Recommended Dilutions: ELISA: 1:2000-1:3000; Dot Blot: 1:200000; Western Blot: 1:1000; Immunofluorescence: 1:200 ChIP: 10 ?l/ChIP)
Type: Primary
Antigen: H3K9me1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human