Anti-PAPPA2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19327
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Pappalysin 2
- Antigen symbol:PAPPA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:60676
- Antigen synonyms:PAPPE|Pappalysin-2|Pregnancy-associated plasma protein A2|PAPP-A2|PAPP-E|PLAC3
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids near N-terminus of human PAPPA2.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody rasied against synthetic peptide of PAPPA2.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 10 ?g/ml
Type: Primary
Antigen: PAPPA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat