Anti-MTR Goat Polyclonal Antibody
Catalog # ABNOPAB6022
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:5-Methyltetrahydrofolate-Homocysteine Methyltransferase
- Antigen symbol:MTR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4548
- Antigen synonyms:cblG|FLJ33168|cobalamin-dependent methionine synthase|MS|MTR|vitamin-B12 dependent methionine synthase|methionine synthase|FLJ45386|5-methyltetrahydrofolate-homocysteine methyltransferase|5-methyltetrahydrofolate-homocysteine methyltransferase 1|FLJ43216
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-VEKWLGPILGYDTD
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human MTR.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of MTR.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 3-5 ?g/ml; ELISA: 1:32000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: MTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human