Order Entry
France
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # 20354-1ML
Supplier:  Biotium
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # 20354-1ML
Supplier:  Biotium

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Conjugation:
    RPE (R-Phycoerythrin)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

R-PE belongs to the family of phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae. R-PE has absorption maxima at 496, 546, and 565 nm respectively and emission at 578 nm, allowing it to be detected in the red channel by flow cytometry using 488 nm excitation. Because of its high extinction coefficient (~1,960,000) and high fluorescence quantum yield (~0.82), R-PE is by far the brightest dye for the 565 nm wavelength region.

Antibodies are supplied at 2 mg/mL in PBS pH 7.4 containing 50% glycerol, 2 mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat