Order Entry
France
ContactUsLinkComponent
 
Anti-gVPLA2 Mouse Monoclonal Antibody [clone: MCL-3G1]
  CAYM160510-100
Anti-gVPLA2 Mouse Monoclonal Antibody [clone: MCL-3G1]
  CAYM160510-100

 

  • Antibody type:
    Primary
  • Antigen symbol:
    GVPLA2
  • Clonality:
    Monoclonal
  • Clone:
    MCL-3G1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human recombinant sPLA2 (type V), W79A mutant
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: gVPLA2
Clonality: Monoclonal
Clone: MCL-3G1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human