Order Entry
France
ContactUsLinkComponent
Anti-CASP1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14F468]
Anti-CASP1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14F468]
Catalog # ROCK200-301-H62
undefined
Anti-CASP1 Mouse Monoclonal Antibody (Unconjugated) [clone: 14F468]
Catalog # ROCK200-301-H62

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    caspase 1, apoptosis-related cysteine peptidase
  • Antigen symbol:
    CASP1
  • Clonality:
    Monoclonal
  • Clone:
    14F468
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    834
  • Antigen synonyms:
    ICE|P45|IL1BC
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Caspase-1 Antibody was produced in mice by repeated immunizations with a synthetic peptide corresponding to amino acids near the c-terminus of the immunogen, human Caspase-1.
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-Caspase-1 antibody is suitable for use in IHC (p) and WB. Expect a band approximately 46kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml; Immunochemistry: titrate to determine concentration

Type: Primary
Antigen: CASP1
Clonality: Monoclonal
Clone: 14F468
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse