Anti-STAT2 Mouse Monoclonal Antibody (Unconjugated) [clone: 19G8.H2.H6]
ROCK200-301-B65
- Antibody type:Primary
- Antigen name:signal transducer and activator of transcription 2, 113kDa
- Antigen symbol:STAT2
- Clonality:Monoclonal
- Clone:19G8.H2.H6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:6773
- Antigen synonyms:P113|ISGF-3|STAT113
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues near the carboxy terminus of mouse STAT2 Protein.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
This Protein-A purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 113 kDa in size corresponding to STAT2 Protein by western blotting.
Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:1000; Immunochemistry: 1:2.5 µg/ml;
Type: Primary
Antigen: STAT2
Clonality: Monoclonal
Clone: 19G8.H2.H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse