Anti-RELA Mouse Monoclonal Antibody (Unconjugated) [clone: 27F9.G4]
Catalog # ROCK200-301-065
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Transcription factor p65
- Antigen symbol:RELA
- Clonality:Monoclonal
- Clone:27F9.G4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:5970
- Antigen synonyms:p65|NFKB3
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:NFkB p65 (Rel A) peptide corresponding to a region near the C-terminus of the human Protein conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This Protein A purified mouse monoclonal antibody is directed against NFkB p65 (Rel A) and a 1:1000 dilution recognizes a 65 kD band by Western blot against HeLa whole cell lysate. Control peptide (100-4165p) is sold separately.
Recommended Dilutions: ELISA: 1:50000-1:100000; Immunofluorescence: 1:5000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:200-1:600;
Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 27F9.G4
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human