Anti-HSF1 Rat Monoclonal Antibody (Unconjugated) [clone: 10H8]
ROCK200-501-F50
- Antibody type:Primary
- Antigen name:Heat shock transcription factor 1
- Antigen symbol:HSF1
- Clonality:Monoclonal
- Clone:10H8
- Conjugation:Unconjugated
- ELISA:Yes
- Gel shift:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Hamster,Rabbit,Monkey,Human,Bovine,Guinea pig,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:3297
- Antigen synonyms:HSTF1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:HSF1 Antibody was produced in rat by repeated immunizations with purified recombinant HSF1 Protein.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Anti-HSF1 Antibody is suitable for WB, IP, IF microscopy, ELISA, and Gel mobility shift assay. Expect a band approximately ~85kDa Protein in unstressed cell lysates, and an ~95kDa Protein in heat shocked cell lysates.
Recommended Dilutions: ELISA: 1:200; Western Blot: 1 µg/ml;
Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone: 10H8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit