Anti-CD75s Mouse Monoclonal Antibody [clone: LN-1]
Catalog # USBI138526
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD75S
- Clonality:Monoclonal
- Clone:LN-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Supernatant
- Form:liquid
- Storage buffer:0,05% sodium azide.
- Storage temperature:–20 °C
- Immunogen:Nuclei from pokeweed mitogen stimulated peripheral blood lymphocytes
- Pk:500 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:25-1:50; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: CD75s
Clonality: Monoclonal
Clone: LN-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human,