Anti-CA19-9 Mouse Monoclonal Antibody [clone: 121SLE]
Catalog # USBI138501
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CA19-9
- Clonality:Monoclonal
- Clone:121SLE
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Supernatant
- Form:liquid
- Storage temperature:–20 °C
- Immunogen:Precipitin lines obtained after immuno-diffusion using MAb 116-NS-19-9 and mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le).
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:100-1:200; Immunofluorescence: 1:25-1:50; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: CA19-9
Clonality: Monoclonal
Clone: 121SLE
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human,