Anti-MHC class I, H-2Kb, Db Mouse Monoclonal Antibody [clone: 28-8-6]
Catalog # USBIM3885-80G-50
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1, H-2Kb, Db
- Clonality:Monoclonal
- Clone:28-8-6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:C3H.SW mouse splenocytes
- Purification:purified by immunoaffinity chromatography
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MHC class I, H-2Kb, Db
Clonality: Monoclonal
Clone: 28-8-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse,