Order Entry
France
ContactUsLinkComponent
 
Anti-Gr-1 Rat Monoclonal Antibody [clone: RB6-8C5]
 
undefined
Anti-Gr-1 Rat Monoclonal Antibody [clone: RB6-8C5]

 

  • Catalog No:
  • USBIG8750-02K-500
  • USBIG8750-02K-50
  • Antigen symbol:
  • GR-1
  • GR-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • RB6-8C5
  • RB6-8C5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Raised against mouse granulocytes
  • Raised against mouse granulocytes
  • Form:
  • liquid
  • liquid
  • Purification:
  • purified by immunoaffinity chromatography
  • purified by immunoaffinity chromatography
  • Concentration:
  • ~0,5 mg/ml
  • ~0,5 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,2, 0,09% sodium azide.
  • PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
  • –20 °C
  • –20 °C

 

 

Frequently Bought Together