Anti-Heat Shock Protein 70, Geodia cydonium Mouse Monoclonal Antibody [clone: IVF5]
Catalog # USBIH1830-92S-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein 70, Geodia cydonium
- Clonality:Monoclonal
- Clone:IVF5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:No
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:HSP70 from Geodia cydonium
- Purification:purified by Protein G chromatography
- Size:10 μg
- Pk:10 µG
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1:1000; Immunochemistry: 1:50; Immunoprecipitation: 1:50
Type: Primary
Antigen: Heat Shock Protein 70, Geodia cydonium
Clonality: Monoclonal
Clone: IVF5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: