Order Entry
France
ContactUsLinkComponent
 
Anti-Bcl 10, Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 6D158]
  USBIB0807-12F
Anti-Bcl 10, Mouse Monoclonal Antibody (KLH (Keyhole Limpet Hemocyanin)) [clone: 6D158]
  USBIB0807-12F

 

  • Antibody type:
    Primary
  • Antigen symbol:
    BCL10
  • Clonality:
    Monoclonal
  • Clone:
    6D158
  • Conjugation:
    KLH (Keyhole Limpet Hemocyanin)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Modification:
    phospho-Ser138
  • Storage buffer:
    PBS, 0,09% sodium azide, PEG, Sucrose and 40% glycerol.
  • Storage temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide corresponding to a portion of Bcl10 phosphorylated at Ser138
  • Purification:
    purified by thiophilic adsorption and size exclusion chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: Bcl 10,
Clonality: Monoclonal
Clone: 6D158
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,