Order Entry
France
ContactUsLinkComponent
Human pulmonary artery endothelial cells (HPAEC)
Human pulmonary artery endothelial cells (HPAEC)
  C-14024
 :  PROMOCELL
 :  
undefined
Human pulmonary artery endothelial cells (HPAEC)
  C-14024
 :  PROMOCELL
 :  
  • Cell Format:
    Cryopreserved
  • Pk:
    2 mL
  • Description:
    Human pulmonary artery endothelial cells (HPAEC) pellet, 1×10⁶ cells in RNAlater®
  • Seq:
    02
Primary Human Pulmonary Artery Endothelial Cells (HPAEC) are isolated from human pulmonary arteries (the main pulmonary artery, left and right branches)
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
Since the pulmonary arteries carry blood from the heart to the lungs, they are the only arteries (other than umbilical arteries) that carry deoxygenated blood.
 : Human Pulmonary Artery Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.