Order Entry
France
ContactUsLinkComponent
Human aortic endothelial cells
Human aortic endothelial cells
Catalog # C-14023
Supplier:  PROMOCELL
CAS Number:  
undefined
Human aortic endothelial cells
Catalog # C-14023
Supplier:  PROMOCELL
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Pk:
    2 mL
  • Description:
    Human aortic endothelial cells (HAoEC) pellet, 1×10⁶ cells in RNAlater®
  • Seq:
    02
Specifications
About this item
Primary Human Aortic Endothelial Cells (HAoEC) are isolated from the human ascending (thoracic) and descending (abdominal) aorta. They are useful for studying vascular diseases such as thrombosis, atherosclerosis and hypertension as well as for stent-graft compatibility testing.
  • Von Willebrand factor (vWF) positive
  • CD31 positive
  • Smooth muscle alpha-actin negative
PromoCell cell pellets are stored in RNAlater®, an aqueous protection solution that rapidly permeates the cells and stabilises cellular DNA, RNA, and proteins and protects them against degradation. This makes the PromoCell cell pellets very useful for a wide range of research applications like gene expression analysis, cell characterisation studies, and protein characterisation experiments. Each Cell Pellet is made from 1 million cells and is dissolved in 200 µl RNAlater®. The Cell Pellets are produced from released cell lots and have the same characteristics, i.e. expression profile or cell type specific antigens.
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies. They are useful for studying vascular diseases such as thrombosis, atherosclerosis, and hypertension as well as for stent-graft compatibility testing.
Ordering information: Human Umbilical Aortic Endothelial Cells can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.