Order Entry
France
ContactUsLinkComponent
Human mesenchymal stem cells from adipose tissue
Human mesenchymal stem cells from adipose tissue
Catalog # C-14092
Supplier:  PROMOCELL
CAS Number:  
undefined
Human mesenchymal stem cells from adipose tissue
Catalog # C-14092
Supplier:  PROMOCELL
CAS Number:  

Specifications

  • Cell Format:
    Pelleted
  • Pk:
    2 mL
  • Description:
    Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT) pellet, 1×106 cells in RNAlater®

Specifications

About this item

Human Mesenchymal Stem Cells from Adipose Tissue (hMSC-AT) are harvested from normal human adipose tissue. The cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes and osteoblasts.

Applications include: Stem cell research, regenerative medicine research, tissue engineering studies, developmental experiments.

PromoCell's hMSC-BM cells show a verified marker expression profile that complies with ISCT* recommendations, providing well characterised cells.

*ISCT (International Society for Cellular Therapy) Cytotherapy (2006) Vol. 8, No. 4, 315-317