Human mesenchymal stem cells from bone marrow
Catalog # C-14090
Supplier: PROMOCELL
CAS Number:
Specifications
- Cell Format:Pelleted
- Pk:2 mL
- Description:Human mesenchymal stem cells from bone marrow (hMSC-BM) pellet, 1×106 cells in RNAlater®
- Seq:03
Specifications
About this item
Mesenchymal stem cells are harvested from normal human bone marrow. All cells are tested for their ability to differentiate in vitro into adipocytes, chondrocytes, osteoblasts and neuronal cells.
PromoCell's hMSC-BM cells show a verified marker expression profile that complies with ISCT* recommendations, providing well characterised cells.
*ISCT (International Society for Cellular Therapy) Cytotherapy (2006) Vol. 8, No. 4, 315-317