Human pericytes from placenta (hPC-PL)
Catalog # C-14095
Supplier: PROMOCELL
CAS Number:
Specifications
- Cell Format:Pelleted
- Pk:2 mL
- Description:Human Pericytes from placenta (hPC-PL) pellet, 1×106 cells in RNAlater®
- Seq:02
Specifications
About this item
Pericytes are multipotent mesenchymal-like cells found in association with small blood vessel walls and are important for angiogenesis, the structural integrity of the microvasculature and blood flow regulation. However, they can also develop into malignant tumours called hemangiopericytomas.
Pericytes have demonstrated the ability to differentiate into adipocytes, osteoblasts, fibroblasts, smooth muscle cells and phagocytes (macrophages).
Ordering information: Human pericytes from human placenta can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.