Human cardiac fibroblasts
Catalog # C-14036
Supplier: PROMOCELL
CAS Number:
Specifications
- Cell Format:Pelleted
- Pk:2 mL
- Description:Human cardiac fibroblasts (HCF) pellet, 1×106 cells in RNAlater®
Specifications
About this item
Primary human cardiac fibroblasts (HCF) are isolated from the ventricles of an adult heart. HCF play a central role in the maintenance of the extracellular matrix in the normal heart and the synthesis of growth factors and cytokines.
Under pathophysiological conditions, cardiac fibroblasts are involved in scar formation after myocardical infarction, cardiac fibrosis and cardiac hypertrophy. HCF can be used to study such processes in vitro.
Delivery: Human Cardiac Fibroblasts can be ordered as cryopreserved or proliferating cells. Cell pellets stored in RNAlater® are also available.