Skeletal muscle cells (SkMC), human
Catalog # C-14060
Supplier: PROMOCELL
CAS Number:
Specifications
- Cell Format:Pelleted
- Pk:2 mL
- Description:Human skeletal muscle cells (SkMC), 1×106 cells, dissolved in 200 μl RNAlater®
- Seq:02
Specifications
About this item
Primary human skeletal muscle cells (SkMC) are isolated from different skeletal muscles (eg. musculus pectoralis major) from adult single donors. They are positive for sarcomeric myosin and negative for smooth muscle specific α-actin.
Primary human skeletal muscle cells (SkMC) are optimal for in vitro muscle studies. They proliferate very well in the mitogen-rich skeletal muscle cell growth medium. Fusion to myotubes with typical multinucleated syncytia can be induced by using the skeletal muscle cell differentiation medium.