Skeletal muscle cell differentiation medium
Catalog # C-23061
Supplier: PROMOCELL
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:Skeletal Muscle Cell Differentiation Medium (ready-to-use), 500 ml
Specifications
About this item
PromoCell Skeletal Muscle Cell Differentiation Medium is a serum-free medium that induces the fusion of myoblasts to myotubes with typical multinucleated syncytia.
The medium is optimised for primary human cells, but can also be used for murine, rat, canine and rabbit skeletal muscle cells.
Delivery: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix.