Mesenchymal stem cell growth medium
Catalog # C-28009
Supplier: PROMOCELL
CAS Number:
Specifications
- Pk:500 mL
- Description:Mesenchymal Stem Cell Growth Medium 2 (ready-to-use), 500 ml
- Seq:01
Specifications
About this item
PromoCell Mesenchymal Stem Cell Growth Medium supports the expansion of multipotent human mesenchymal stem cells (MSC) without inducing early senescence and differentiation as observed in standard culture media.
Delivery: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix.