Osteoblast growth medium
Supplier: PROMOCELL
Certificates
About this item
PromoCell Osteoblast Growth Medium was developed to establish and maintain human osteoblast cultures. The foetal calf serum in the medium is strictly pretested for optimal performance.
The media are optimised for primary human cells but can also be used for bovine, murine and rat osteoblasts, as well as osteoblast cell lines.
Delivery: The medium (ready-to-use) consists of 1× 500 ml bottle basal medium and 1× vial of SupplementMix. Basal medium also available separately.