Anti-UTS2 Rabbit Polyclonal Antibody
USBIU2800-03B
- Antibody type:Primary
- Antigen name:Urotensin 2
- Antigen symbol:UTS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: UTS2 (urotensin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human