Order Entry
France
ContactUsLinkComponent
Anti-TBX4 Rabbit Polyclonal Antibody
Anti-TBX4 Rabbit Polyclonal Antibody
Catalog # USBIT1848-26T
Supplier:  US Biological
Anti-TBX4 Rabbit Polyclonal Antibody
Catalog # USBIT1848-26T
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    T-box 4
  • Antigen symbol:
    TBX4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Form:
    liquid
  • Amino acid number:
    529 to 545
  • Storage buffer:
    PBS, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TBX4 (T-box 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human