Anti-SUZ12 Mouse Monoclonal Antibody [clone: 8H195]
Catalog # USBIS8995-02
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:SUZ12 polycomb repressive complex 2 subunit
- Antigen symbol:SUZ12
- Clonality:Monoclonal
- Clone:8H195
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Amino acid number:131 to 305
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1-2?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: SUZ12 (SUZ12 polycomb repressive complex 2 subunit)
Clonality: Monoclonal
Clone: 8H195
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human