Anti-Propoxyphene Sheep Polyclonal Antibody
USBIP9052-50
- Antibody type:Primary
- Antigen symbol:Propoxyphene
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Recommended dilution: ELISA: 0. 625 ?g/ml
Type: Primary
Antigen: Propoxyphene
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: