Order Entry
France
ContactUsLinkComponent
 
Anti-PIP4K2A Mouse Monoclonal Antibody [clone: 3A3]
  USBIP4071-77A
undefined
Anti-PIP4K2A Mouse Monoclonal Antibody [clone: 3A3]
  USBIP4071-77A

 

  • Antibody type:
    Primary
  • Antigen name:
    phosphatidylinositol-5-phosphate 4-kinase, type II, alpha
  • Antigen symbol:
    PIP4K2A
  • Clonality:
    Monoclonal
  • Clone:
    3A3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    304 to 366
  • Storage buffer:
    PBS, pH 7,2.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PIP4K2A (phosphatidylinositol-5-phosphate 4-kinase, type II, alpha)
Clonality: monoclonal
Clone: 3A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human