Order Entry
France
ContactUsLinkComponent
Anti-PIP5K1A Rabbit Polyclonal Antibody (Biotin)
Anti-PIP5K1A Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIP4071-76A-BIOT
Supplier:  US Biological
undefined
Anti-PIP5K1A Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIP4071-76A-BIOT
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    phosphatidylinositol-4-phosphate 5-kinase, type I, alpha
  • Antigen symbol:
    PIP5K1A
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: PIP5K1A (phosphatidylinositol-4-phosphate 5-kinase, type I, alpha)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human