Anti-HECW2 Goat Polyclonal Antibody
USBIN0581-15B
- Antibody type:Primary
- Antigen name:HECT, C2 and WW domain containing E3 ubiquitin Protein ligase 2
- Antigen symbol:HECW2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Bovine,Rat,Mouse
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:32000
Type: Primary
Antigen: HECW2 (HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat