Anti-MHC Class 1, H2, Dd Mouse Monoclonal Antibody (Biotin) [clone: 5K47]
Catalog # USBIM3885-63B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1, H2, Dd
- Clonality:Monoclonal
- Clone:5K47
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: MHC Class 1, H2, Dd
Clonality: monoclonal
Clone: 5K47
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse