Anti-Melanoma Discriminating Antigen Mouse Monoclonal Antibody [clone: PAL-M1]
USBIM2869-40
- Antibody type:Primary
- Antigen name:Melanoma Discriminating Antigen
- Clonality:Monoclonal
- Clone:PAL-M1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Environmentally Preferable:
- Format:Supernatant
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Immunochemistry: 1:5-1:25
Type: Primary
Antigen: Melanoma Discriminating Antigen
Clonality: monoclonal
Clone: PAL-M1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: