Anti-LEP Mouse Monoclonal Antibody [clone: 7C216]
Catalog # USBIL1670-08Q
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:leptin
- Antigen symbol:LEP
- Clonality:Monoclonal
- Clone:7C216
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:131 to 145
- Storage buffer:PBS,
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LEP (leptin)
Clonality: monoclonal
Clone: 7C216
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human