Anti-LEP Rabbit Polyclonal Antibody
Catalog # USBIL1670-07K
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:leptin
- Antigen symbol:LEP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:0,5XPBS, pH 7,4.
- Storage temperature:Cool
- Concentration:~1 mg/ml (prior to lyophilization)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5 ?g/ml; Western Blot: 0.1-0.2?g/ml
Type: Primary
Antigen: LEP (leptin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat