Anti-Heparan Sulfate Mouse Monoclonal Antibody [clone: 8.S.087]
Catalog # USBIH1890
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Heparan Sulfate
- Clonality:Monoclonal
- Clone:8.S.087
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by thiophilic affinity chromatography using Prosep Thiosorb M
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:70-1:140; Immunochemistry: 1:35-1:70
Type: Primary
Antigen: Heparan Sulfate
Clonality: monoclonal
Clone: 8.S.087
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human