Anti-FPR3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # USBIF6011-01A-FITC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Formyl Peptide Receptor 3
- Antigen symbol:FPR3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:313 to 342
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Flow Cytometry: 1:10-1:50; Immunochemistry: 1:50-1:100; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: FPR3 (formyl peptide receptor 3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human