Anti-FOLR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11C18]
USBIF5750-16B
- Antibody type:Primary
- Antigen name:Folate receptor 1
- Antigen symbol:FOLR1
- Clonality:Monoclonal
- Clone:11C18
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Amino acid number:25 to 233
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,05 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Do not freeze!
Type: Primary
Antigen: FOLR1 (folate receptor 1 (adult))
Clonality: monoclonal
Clone: 11C18
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human