Anti-Fluorescein Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Référence USBIF5200-15A
Fournisseur: US Biological
Spécifications
- Antibody type:Tag
- Antigen name:Fluorescein
- Antigen symbol:FITC
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Goat
- Reactivity:Goat
- Western blot:Yes
- Form:liquid
- Storage buffer:0,05M tris chloride, 0,15M sodium chloride,0,001M magnesium chloride, 0,0001M zinc chloride, 50% glycerol
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by immunoaffinity chromatography using Fluorescein coupled to agarose beads followed by solid phase adsorption
- Size:1 mg
- Pk:1 mg
Spécifications
À propos de cet article
Recommended dilution: ELISA: 1:1000 to 1:5000
Type: Tag
Antigen: Fluorescein (fluorescein)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Goat