Order Entry
France
ContactUsLinkComponent
 
Anti-FGFR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBIF4305-08B-PE
Supplier:  US Biological
undefined
Anti-FGFR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBIF4305-08B-PE
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor receptor 1
  • Antigen symbol:
    FGFR1
  • Clonality:
    Polyclonal
  • Conjugation:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Tyr307
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Dot Blot, Fluoresence ELISA
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000 Dot Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: FGFR1 (fibroblast growth factor receptor 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human