Anti-Fibrinogen Goat Polyclonal Antibody (Biotin)
Catalog # USBIF4200-07B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibrinogen
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 10mg/ml BSA, 0,01% sodium azide, 40% glycerol
- Storage temperature:Cool
- Concentration:~10 mg/ml
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by delipidation, salt fractionation and ion exchange chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:50000-1:250000
Type: Primary
Antigen: Fibrinogen (fibrinogen)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human