Anti-F9 Mouse monoclonal antibody [clone: 8.H.1]
Catalog # USBIF0017
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:coagulation factor IX
- Antigen symbol:F9
- Clonality:Monoclonal
- Clone:8.H.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:50% glycerol
- Storage temperature:Cool
- Concentration:~10 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 5-10 ?g/ml; Immunochemistry: 100ng/ml
Type: Primary
Antigen: F9 (coagulation factor IX)
Clonality: monoclonal
Clone: 8.H.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human