Anti-Epidermal Growth Factor-Like Module-Containing Mucin-Like Receptor 3 Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10C111]
USBIE3375-79C4
- Antibody type:Primary
- Antigen name:Epidermal Growth Factor-Like Module-Containing Mucin-Like Receptor 3
- Clonality:Monoclonal
- Clone:10C111
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide, 1% BSA.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Recommended dilution: Flow Cytometry:1:5;
Do not freeze!
Type: Primary
Antigen: Epidermal Growth Factor-Like Module-Containing Mucin-Like Receptor 3
Clonality: monoclonal
Clone: 10C111
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human