Anti-TRDMT1 Rabbit Polyclonal Antibody (Biotin)
USBID3931-62D-BIOT
- Antibody type:Primary
- Antigen name:tRNA aspartic acid methyltransferase 1
- Antigen symbol:TRDMT1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse