Anti-CBS Mouse Monoclonal Antibody [clone: 6B8]
Catalog # USBIC8948-50E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cystathionine-beta-synthase
- Antigen symbol:CBS
- Clonality:Monoclonal
- Clone:6B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 101
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CBS (cystathionine-beta-synthase)
Clonality: Monoclonal
Clone: 6B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human