Anti-Complement C5a des-Arg Mouse Monoclonal Antibody [clone: 6A535]
Catalog # USBIC7850-24H
Fournisseur: US Biological
Spécifications
- Antibody type:Primary
- Antigen name:Complement C5a des-Arg
- Clonality:Monoclonal
- Clone:6A535
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:liquid
- Amino acid number:65 to 73
- Storage buffer:PBS, 0,1% BSA.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Spécifications
À propos de cet article
Recommended dilution: Western Blot: 1:50; Immunochemistry: 1 ?g/ml
Type: Primary
Antigen: Complement C5a des-Arg
Clonality: Monoclonal
Clone: 6A535
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human