Anti-CD178 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11n69]
Catalog # USBIC2548-91P1
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CD178
- Clonality:Monoclonal
- Clone:11n69
- Conjugation:APC (Allophycocyanin)
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Form:liquid
- Amino acid number:132 to 279
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: CD178
Clonality: Monoclonal
Clone: 11n69
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse