Anti-ITGA2B Mouse Monoclonal Antibody [clone: 5F36]
Catalog # USBIC2394-04M
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 2 Beta
- Antigen symbol:ITGA2B
- Clonality:Monoclonal
- Clone:5F36
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG3
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: Monoclonal
Clone: 5F36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human