Order Entry
France
ContactUsLinkComponent
Anti-CD40 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5K254]
Anti-CD40 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5K254]
Catalog # USBIC2392-17E
Supplier:  US Biological
Anti-CD40 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5K254]
Catalog # USBIC2392-17E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD40 molecule, TNF receptor superfamily member 5
  • Antigen symbol:
    CD40
  • Clonality:
    Monoclonal
  • Clone:
    5K254
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide and a stabilizing agent
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: Monoclonal
Clone: 5K254
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse